Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-fluoro-6-methylaniline 97%

Supplier: AMBEED, INC
Warning

A179134-5G A179134-100G A179134-10G A179134-500G A179134-1G A179134-5G A179134-25G
103807-400EA 5 USD InStock
103807-400 103845-742 76769-502 76769-504 76769-506 77083-192 77083-194 77301-596
2-Bromo-4-fluoro-6-methylaniline 97%
2-Bromo-4-fluoro-6-methylaniline
Precaución: for Research Use Only


Formula: C₇H₇BrFN
Boiling Pt: 248.9 °C (760 mm Hg)
Melting Pt: 32…36 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01861196
CAS Number: 202865-77-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty