Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-fluorobenzonitrile 98%

Supplier: AMBEED, INC
Warning

A229574-5G A229574-10G A229574-25G A229574-5G A229574-100G
103850-384EA 8 USD InStock
103850-384 76771-102 77089-232 77089-234 77626-210
2-Bromo-5-fluorobenzonitrile 98%
2-Bromo-5-fluorobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₃BrFN
MW: 200.01 g/mol
Boiling Pt: 255
Melting Pt: 92…95 °C
Flash Pt: 105.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00142875
CAS Number: 57381-39-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty