Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-methoxy-4-methylaniline 98%

Supplier: AMBEED, INC
Warning

A160083-1G A160083-5G A160083-250MG A160083-1G A160083-5G
103805-330EA 81 USD InStock
103805-330 103843-560 76997-764 77319-476 77319-478
2-Bromo-5-methoxy-4-methylaniline 98%
2-Bromo-5-methoxy-4-methylaniline
Precaución: for Research Use Only


Formula: C₈H₁₀BrNO
Melting Pt: 75…77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11841053
CAS Number: 532440-88-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty