2-Bromo-5-nitrothiophene 97%

Supplier: AMBEED, INC
Warning

A132067-1G A132067-10G A132067-250MG A132067-1G A132067-25G A132067-5G
103801-684EA 6 USD
103801-684 76811-252 76811-254 77314-234 77314-236 77314-238
2-Bromo-5-nitrothiophene 97%
2-Bromo-5-nitrothiophene

Precaución: for Research Use Only

Formula: C₄H₂BrNO₂S
MW: 208.04
Melting Pt: 44…48 °C
Flash Pt: 235 °C
Storage Temperature: Ambient
MDL Number: MFCD00022493
CAS Number: 13195-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR