Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3-fluoro-5-hydroxypyridine 97%

Supplier: AMBEED, INC
Warning

A184061-1G A184061-5G A184061-10G A184061-250MG A184061-1G A184061-5G
103815-708EA 45 USD InStock
103815-708 103846-268 76974-542 76974-544 77083-878 77083-880
2-Chloro-3-fluoro-5-hydroxypyridine 97%
2-Chloro-3-fluoro-5-hydroxypyridine
Precaución: for Research Use Only


Formula: C₆H₄ClFO
Boiling Pt: 347 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03411573
CAS Number: 870062-76-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty