Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4,6-difluoroaniline 97%

Supplier: AMBEED, INC
Warning

A174031-1G A174031-5G A174031-100G A174031-10G A174031-1G A174031-25G
103806-904EA 9 USD InStock
103806-904 103845-204 77096-570 77096-572 77301-308 77301-310
2-Chloro-4,6-difluoroaniline 97%
2-Chloro-4,6-difluoroaniline
Precaución: for Research Use Only


Formula: C₆H₄ClF₂N
Boiling Pt: 178.6 °C (760 mm Hg)
Melting Pt: 28 °C
Flash Pt: 82.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00142892
CAS Number: 36556-56-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty