Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-methylpyrimidine 98%

Supplier: AMBEED, INC
Warning

A174952-5G A174952-1G A174952-25G A174952-5G A174952-100G A174952-10G
103806-992EA 5 USD InStock
103806-992 103845-292 77082-604 77082-606 77082-608 77111-168 77111-170
2-Chloro-5-methylpyrimidine 98%
2-Chloro-5-methylpyrimidine
Precaución: for Research Use Only


Formula: C₅H₅ClN₂
Boiling Pt: 239.2 °C (760 mm Hg)
Melting Pt: 89…92 °C
Flash Pt: 121.5 °C
Storage Temperature: Ambient
MDL Number: MFCD09260903
CAS Number: 22536-61-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty