Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxycyclopent-2-en-1-one 97%

Supplier: AMBEED, INC
Warning

A169331-1G A169331-5G A169331-100MG A169331-10G A169331-250MG A169331-1G A169331-5G
103806-374EA 48 USD InStock
103806-374 103844-628 76973-912 76973-914 76973-916 77081-704 77081-706
2-Hydroxycyclopent-2-en-1-one 97%
2-Hydroxycyclopent-2-en-1-one
Precaución: for Research Use Only


Formula: C₅H₆O₂
Boiling Pt: 244.8 °C (760 mm Hg)
Melting Pt: 52…53 °C
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16251523
CAS Number: 10493-98-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty