Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxyethanesulfonic acid 80% (w/w) in water 80% (w/w)

Supplier: AMBEED, INC
Danger

A638786-100G A638786-10G A638786-25G
103840-326EA 8 USD InStock
103840-326 76806-294 76806-296 77308-036
2-Hydroxyethanesulfonic acid 80% (w/w) in water 80% (w/w)
2-Hydroxyethanesulfonic acid
Precaución: for Research Use Only


Formula: C₂H₆O₄S
Storage Temperature: Refrigerator
MDL Number: MFCD00242599
CAS Number: 107-36-8
UN: 3265
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty