Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodobenzonitrile 98%

Supplier: AMBEED, INC
Warning

A502312-5G A502312-100G A502312-10G A502312-500G A502312-25G
103839-388EA 7 USD InStock
103839-388 77279-692 77279-694 77279-696 77307-622
2-Iodobenzonitrile 98%
2-Iodobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₄IN
Boiling Pt: 283.1 °C (760 mm Hg)
Melting Pt: 52…54 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00079761
CAS Number: 4387-36-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty