Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Hydroxytryptophol 98%

Supplier: AMBEED, INC
Warning

A209297-1G A209297-250MG A209297-100MG
103817-932EA 262 USD InStock
103817-932 77626-060 77877-657
5-Hydroxytryptophol 98%
5-Hydroxytryptophol
Precaución: Keep in a dark place, inert atmosphere, store in freezer, under -20 °C


Formula: C₁₀H₁₁NO₂
Melting Pt: 111…113 °C
Storage Temperature: Freezer
MDL Number: MFCD00047197
CAS Number: 154-02-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty