Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl(3-pyridyl)borane 97%

Supplier: AMBEED, INC
Warning

A163979-1G A163979-5G A163979-100G A163979-10G A163979-25G A163979-1G A163979-5G
103805-796EA 6 USD InStock
103805-796 103844-062 76799-124 76799-126 77300-688 77320-188 77320-190
Diethyl(3-pyridyl)borane 97%
Diethyl(3-pyridyl)borane
Precaución: for Research Use Only


Formula: C₉H₁₄BN
Boiling Pt: 205.9 °C (760 mm Hg)
Melting Pt: 172…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00012348
CAS Number: 89878-14-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty