3,4-Dimethoxythiophene 97%

Supplier: AMBEED, INC
Warning

A121854-1G A121854-10G A121854-25G A121854-1G A121854-5G A121854-500G
103800-146EA 5 USD
103800-146 76766-540 76766-542 77312-276 77312-278 77876-885
3,4-Dimethoxythiophene 97%
3,4-Dimethoxythiophene

Precaución: For research use only.

Formula: C6H8O2S
MW: 144.19 g/mol
Boiling Pt: 184.4 °C (760 mm Hg)
Flash Pt: 106.7 °C
Storage Temperature: Freezer
MDL Number: MFCD01096546
CAS Number: 51792-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR