(+/-)-Dehydrolinalool 95%

Supplier: AMBEED, INC
Warning

A108516-100G A108516-10G A108516-25G A108516-5G
103823-142EA 17 USD
103823-142 77270-698 77270-700 77270-702 77309-988
(+/-)-Dehydrolinalool 95%
(+/-)-Dehydrolinalool

Precaución: for Research Use Only

Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 228.6 °C (760 mm Hg)
Flash Pt: 167 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00129075
CAS Number: 29171-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR