Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-o-toluic acid 95%

Supplier: AMBEED, INC
Warning

A219284-5G A219284-25G A219284-100G A219284-500G A219284-5G
103809-546EA 346 USD InStock
103809-546 103849-652 77088-206 77249-342 77249-344 77303-524
3-Amino-o-toluic acid 95%
3-Amino-o-toluic acid
Precaución: for Research Use Only


Formula: C₈H₉NO₂
Boiling Pt: 347.3 °C (760 mm Hg)
Melting Pt: 178…181 °C
Flash Pt: 163.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00075026
CAS Number: 52130-17-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty