Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-5-bromo-4-methylpyridin-2-ol 97%

Supplier: AMBEED, INC
Warning

A217280-1G A217280-5G A217280-100MG A217280-250MG A217280-1G A217280-5G
103818-612EA 17 USD InStock
103818-612 103849-344 76986-930 76986-932 77087-958 77087-960
3-Amino-5-bromo-4-methylpyridin-2-ol 97%
3-Amino-5-bromo-4-methylpyridin-2-ol
Precaución: for Research Use Only


Formula: C₆H₇BrN₂O
Boiling Pt: 288.5±40.0 °C (760 mm Hg)
Flash Pt: 128.3 °C
Storage Temperature: Ambient
MDL Number: MFCD08436400
CAS Number: 889943-27-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty