Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1-Dioxidotetrahydrothien-3-ylamine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A122786-1G A122786-100MG A122786-10G A122786-250MG A122786-5G
103800-282EA 28 USD InStock
103800-282 77271-606 77271-608 77271-610 77312-400
1,1-Dioxidotetrahydrothien-3-ylamine hydrochloride 97%
1,1-Dioxidotetrahydrothien-3-ylamine hydrochloride
Precaución: for Research Use Only


Formula: C₄H₁₀ClNO₂S
Melting Pt: 208 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00456584
CAS Number: 51642-03-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty