Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-(trifluoromethyl)pyrazole 98%

Supplier: AMBEED, INC
Warning

A116418-1G A116418-100MG A116418-1G A116418-5G
103798-732EA 107 USD InStock
103798-732 103824-598 76810-210 77297-462 77297-464
3-Bromo-5-(trifluoromethyl)pyrazole 98%
3-Bromo-5-(trifluoromethyl)pyrazole
Precaución: for Research Use Only


Formula: C₄H₂BrF₃N₂
MW: 214.971 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12755885
CAS Number: 93608-11-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty