Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-fluorophenylacetonitrile 98%

Supplier: AMBEED, INC
Warning

A473801-1G A473801-25G A473801-5G A473801-10G
103828-056EA 15 USD InStock
103828-056 103839-130 76843-050 76843-052 76843-054 77004-504
3-Chloro-2-fluorophenylacetonitrile 98%
3-Chloro-2-fluorophenylacetonitrile
Precaución: for Research Use Only


Formula: C₈H₅ClFN
MW: 169.5800 g/mol
Boiling Pt: 256.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01631552
CAS Number: 261762-98-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty