Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-hydroxybenzonitrile 97%

Supplier: AMBEED, INC
Warning

A664333-5G A664333-25G A664333-100G A664333-10G A664333-1G
103840-460EA 13 USD InStock
103840-460 76844-654 76980-880 76980-882 77308-078
3-Fluoro-5-hydroxybenzonitrile 97%
3-Fluoro-5-hydroxybenzonitrile
Precaución: For research use only


Formula: C₇H₄FNO
MW: 137.1112 g/mol
Melting Pt: 138…139 °C
Storage Temperature: Ambient
MDL Number: MFCD09972133
CAS Number: 473923-95-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty