Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxy-2,2-dimethylpropanenitrile 97%

Supplier: AMBEED, INC
Warning

A114582-1G A114582-100MG A114582-250MG A114582-1G A114582-5G
103798-424EA 88 USD InStock
103798-424 103824-264 76995-430 76995-432 77311-080 77311-082
3-Hydroxy-2,2-dimethylpropanenitrile 97%
3-Hydroxy-2,2-dimethylpropanenitrile
Precaución: for Research Use Only


Formula: C₅H₉NO
Storage Temperature: Refrigerator
MDL Number: MFCD12028479
CAS Number: 19295-57-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty