Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Biphenyl isothiocyanate 97%

Supplier: AMBEED, INC
Warning

A123473-1G A123473-250MG A123473-1G
103800-410EA 307 USD InStock
103800-410 77094-588 77312-528
3-Biphenyl isothiocyanate 97%
3-Biphenyl isothiocyanate
Precaución: for Research Use Only


Formula: C₁₃H₉NS
MW: 211.29 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09025658
CAS Number: 1510-25-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty