Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A219175-1G A219175-5G A219175-100MG A219175-250MG A219175-1G A219175-5G
103818-766EA 12 USD InStock
103818-766 103849-640 77030-740 77030-742 77088-182 77088-184
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine hydrochloride 95%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₂₀BNO₂·ClH
Storage Temperature: Freezer
MDL Number: MFCD11506076
CAS Number: 1121057-75-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty