Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Benzyloxy)-3-hydroxybenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A100482-1G A100482-5G A100482-100MG A100482-250MG A100482-1G
103795-468EA 54 USD InStock
103795-468 103821-162 76780-370 76780-372 77296-714
4-(Benzyloxy)-3-hydroxybenzaldehyde 97%
4-(Benzyloxy)-3-hydroxybenzaldehyde
Precaución: for Research Use Only


Formula: C₁₄H₁₂O₃
Boiling Pt: 403.3 °C (760 mm Hg)
Flash Pt: 155.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03840812
CAS Number: 4049-39-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty