Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,7-Dibromo[1,2,5]thiadiazolo[3,4-c]pyridine 96%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A318623-1G A318623-5G A318623-1G A318623-250MG
103825-176EA 63 USD InStock
103825-176 103836-110 77306-182 77683-947
4,7-Dibromo[1,2,5]thiadiazolo[3,4-c]pyridine 96%, Ambeed.Inc
4,7-Dibromo[1,2,5]thiadiazolo[3,4-c]pyridine
Precaución: For research use only.


Formula: C₅HBr₂N₃S
Boiling Pt: 347.8 °C (760 mm Hg)
Melting Pt: 144…149 °C
Flash Pt: 164.1 °C
Storage Temperature: Freezer
MDL Number: MFCD23703147
CAS Number: 333432-27-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty