Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Phenylbenzophenone 98%

Supplier: AMBEED, INC
Warning

A111995-1000G A111995-100G A111995-25G A111995-500G A111995-5G
103823-762EA 11 USD InStock
103823-762 77025-796 77025-798 77025-800 77025-802 77284-200
4-Phenylbenzophenone 98%
4-Phenylbenzophenone
Precaución: for Research Use Only


Formula: C₁₉H₁₄O
Boiling Pt: 419.1 °C (760 mm Hg)
Melting Pt: 99…101 °C
Flash Pt: 190.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00003079
CAS Number: 2128-93-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty