Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromoimidazole 98%

Supplier: AMBEED, INC
Danger

A166871-1G A166871-5G A166871-10G A166871-1G A166871-25G A166871-5G
103806-106EA 9 USD InStock
103806-106 103844-368 76973-824 77320-646 77320-648 77320-650
4-Bromoimidazole 98%
4-Bromoimidazole
Precaución: for Research Use Only


Formula: C₃H₃BrN₂
Melting Pt: 131…135 °C
Flash Pt: 150.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00047021
CAS Number: 2302-25-2
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty