Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-1H-pyrazolo[3,4-c]pyridine 97%

Supplier: AMBEED, INC
Danger

A189141-1G A189141-5G A189141-100MG A189141-250MG A189141-5G A189141-1G
103816-160EA 54 USD InStock
103816-160 103846-752 76974-788 76974-790 77084-506 77302-168
4-Bromo-1H-pyrazolo[3,4-c]pyridine 97%
4-Bromo-1H-pyrazolo[3,4-c]pyridine
Precaución: for Research Use Only


Formula: C₆H₄BrN₃
Boiling Pt: 357.5 °C (760 mm Hg)
Flash Pt: 170.029 °C
Storage Temperature: Ambient
MDL Number: MFCD11109831
CAS Number: 1032943-43-3
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty