Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-6-chloro-1H-indole 95%

Supplier: AMBEED, INC
Warning

A435005-1G A435005-5G A435005-100MG A435005-250MG A435005-1G A435005-5G
103827-586EA 77 USD InStock
103827-586 103838-652 76990-552 76990-554 77307-284 77307-286
4-Bromo-6-chloro-1H-indole 95%
4-Bromo-6-chloro-1H-indole
Precaución: for Research Use Only


Formula: C₈H₅BrClN
Boiling Pt: 348.1±22.0 °C (760 mm Hg)
Flash Pt: 164.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07781364
CAS Number: 885519-23-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty