Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-6-methoxyquinoline 97%

Supplier: AMBEED, INC
Warning

A155888-1G A155888-5G A155888-10G A155888-250MG A155888-1G A155888-5G
103804-812EA 30 USD InStock
103804-812 103843-014 76798-786 76798-788 77318-756 77318-758
4-Chloro-6-methoxyquinoline 97%
4-Chloro-6-methoxyquinoline
Precaución: for Research Use Only


Formula: C₁₀H₈ClNO
Boiling Pt: 299.9 °C (760 mm Hg)
Flash Pt: 135.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00024007
CAS Number: 4295-04-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty