Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chlorobenzoic anhydride 97%

Supplier: AMBEED, INC
Warning

A412141-10G A412141-250MG A412141-1G A412141-5G A412141-25G
103827-244EA 27 USD InStock
103827-244 103838-290 76725-178 76725-180 76841-832 76841-834 77307-156
4-Chlorobenzoic anhydride 97%
4-Chlorobenzoic anhydride
Precaución: for Research Use Only


Formula: C₁₄H₈Cl₂O₃
Boiling Pt: 439.0±30.0 °C (760 mm Hg)
Melting Pt: 66.5…70.0 °C
Flash Pt: 182.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00054415
CAS Number: 790-41-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty