Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethynylbenzoic acid 98%

Supplier: AMBEED, INC
Warning

A141073-1G A141073-100G A141073-10G A141073-250MG A141073-1G A141073-25G A141073-5G
103802-948EA 7 USD InStock
103802-948 77010-344 77010-346 77010-348 77316-074 77316-076 77316-078
4-Ethynylbenzoic acid 98%
4-Ethynylbenzoic acid
Precaución: for Research Use Only


Formula: C₉H₆O₂
Melting Pt: 223…227 °C
Flash Pt: 126.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00168819
CAS Number: 10602-00-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty