4-Fluoro-2-iodobenzoic acid 98%

Supplier: AMBEED, INC
Danger

A257899-1G A257899-5G A257899-100MG A257899-10G A257899-250MG A257899-1G A257899-25G A257899-5G
103812-344EA 8 USD
103812-344 103852-218 76976-848 76976-850 76976-852 77091-740 77091-742 77091-744
4-Fluoro-2-iodobenzoic acid 98%
4-Fluoro-2-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄FIO₂
MW: 266.01 g/mol
Boiling Pt: 309.3 °C (760 mm Hg)
Melting Pt: 143…147 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04108278
CAS Number: 56096-89-0
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR