Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-1-naphthaleneboronic acid 98%

Supplier: AMBEED, INC
Warning

A181307-1G A181307-5G A181307-100MG A181307-10G A181307-250MG
103807-596EA 16 USD InStock
103807-596 103845-964 77287-216 77287-218 77287-220
4-Fluoro-1-naphthaleneboronic acid 98%
4-Fluoro-1-naphthaleneboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₇BBrFO₂
Boiling Pt: 374 °C (760 mm Hg)
Melting Pt: 240…242 °C
Flash Pt: 180 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039221
CAS Number: 182344-25-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty