Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[d]thiazole 97%

Supplier: AMBEED, INC
Warning

A150133-1G A150133-5G A150133-100MG A150133-1G A150133-250MG
103804-132EA 36 USD InStock
103804-132 103842-266 76871-582 77317-772 77671-112
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[d]thiazole 97%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[d]thiazole
Precaución: for Research Use Only


Formula: C₁₃H₁₆BNO₂S
Boiling Pt: 380.6±15.0 °C (760 mm Hg)
Flash Pt: 184 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09260439
CAS Number: 1073354-91-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty