Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,6-Dichloropicolinic acid 98%

Supplier: AMBEED, INC
Warning

A616215-1G A616215-25G A616215-5G A616215-10G A616215-250MG
103840-192EA 103 USD InStock
103840-192 76844-358 76844-360 76844-362 76980-584 76980-586
5,6-Dichloropicolinic acid 98%
5,6-Dichloropicolinic acid
Precaución: For research use only


Formula: C₆H₃Cl₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD13185546
CAS Number: 88912-24-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty