Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-ethynylpyridine 98%

Supplier: AMBEED, INC
Warning

A149507-1G A149507-5G A149507-100MG A149507-250MG A149507-1G A149507-5G
103804-050EA 29 USD InStock
103804-050 103842-166 77150-328 77150-330 77317-668 77317-670
5-Bromo-2-ethynylpyridine 98%
5-Bromo-2-ethynylpyridine
Precaución: for Research Use Only


Formula: C₇H₄BrN
Boiling Pt: 212.1 °C (760 mm Hg)
Melting Pt: 84…86 °C
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095276
CAS Number: 111770-86-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty