Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-3-fluorobenzene-1,2-diol 97%

Supplier: AMBEED, INC
Warning

A121623-1G A121623-100MG A121623-250MG A121623-1G
103800-096EA 55 USD InStock
103800-096 76822-458 76822-460 77312-222
5-Bromo-3-fluorobenzene-1,2-diol 97%
5-Bromo-3-fluorobenzene-1,2-diol
Precaución: for Research Use Only


Formula: C₆H₄BrFO₂
Boiling Pt: 263.2±35.0 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD19441229
CAS Number: 876861-29-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty