Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-3-iodo-1H-indazole 98%

Supplier: AMBEED, INC
Warning

A849304-10G A849304-250MG A849304-1G A849304-5G
103841-230EA 20 USD InStock
103841-230 76776-728 76776-730 76845-474 76845-476
5-Bromo-3-iodo-1H-indazole 98%
5-Bromo-3-iodo-1H-indazole
Precaución: for Research Use Only


Formula: C₇H₄BrIN₂
Storage Temperature: Refrigerator
MDL Number: MFCD07781637
CAS Number: 459133-66-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty