Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5H-Pyrido[3,2-b]indole 95%

Supplier: AMBEED, INC
Danger

A531413-1G A531413-5G A531413-100MG A531413-250MG A531413-1G A531413-5G
103828-568EA 43 USD InStock
103828-568 103839-680 76991-524 76991-526 77307-742 77307-744
5H-Pyrido[3,2-b]indole 95%
5H-Pyrido[3,2-b]indole
Precaución: for Research Use Only


Formula: C₁₁H₈N₂
Boiling Pt: 372.9 °C (760 mm Hg)
Flash Pt: 171.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13178683
CAS Number: 245-08-9
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty