Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Hydroxy-L-tryptophan 97%

Supplier: AMBEED, INC
Danger

A102374-5G A102374-100MG A102374-10G A102374-25G A102374-5G
103822-060EA 8 USD InStock
103822-060 77093-568 77093-570 77093-572 77308-948
5-Hydroxy-L-tryptophan 97%
5-Hydroxy-L-tryptophan
Precaución: for Research Use Only


Formula: C₁₁H₁₂N₂O₃
Boiling Pt: 520.6 °C (760 mm Hg)
Melting Pt: 270 °C
Flash Pt: 268.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064341
CAS Number: 4350-09-8
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty