Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methoxy-1H-pyrrolo[2,3-c]pyridine 95%

Supplier: AMBEED, INC
Warning

A159786-1G A159786-5G A159786-1G
103805-288EA 19 USD InStock
103805-288 77011-172 77300-418
5-Methoxy-1H-pyrrolo[2,3-c]pyridine 95%
5-Methoxy-1H-pyrrolo[2,3-c]pyridine
Precaución: for Research Use Only


Formula: C₈H₈N₂O
Boiling Pt: 304.1 °C (760 mm Hg)
Melting Pt: 122…127 °C
Flash Pt: 104.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09038907
CAS Number: 17288-53-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty