Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methoxynicotinic acid 97%

Supplier: AMBEED, INC
Warning

A265626-1G A265626-5G A265626-1G A265626-10G A265626-250MG
103812-740EA 16 USD InStock
103812-740 103852-634 77092-334 77140-848 77140-850
5-Methoxynicotinic acid 97%
5-Methoxynicotinic acid
Precaución: for Research Use Only


Formula: C7H7NO3
Melting Pt: 127…131 °C
Storage Temperature: Ambient
MDL Number: MFCD01603648
CAS Number: 20826-03-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty