Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%

Supplier: AMBEED, INC
Warning

A243272-1G A243272-100MG A243272-250MG A243272-1G
103811-512EA 469 USD InStock
103811-512 76826-772 76826-774 77090-532
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,4-tetrahydroisoquinoline hydrochloride
Precaución: for Research Use Only


Formula: C₁₅H₂₃BClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD30342161
CAS Number: 2828446-91-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty