Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-1-tert-Butyl-2-methyl pyrrolidine-1,2-dicarboxylate 97%

Supplier: AMBEED, INC
Warning

A161837-5G A161837-5G A161837-25G A161837-100G A161837-10G
103843-784EA 7 USD InStock
103843-784 77300-560 77319-798 77611-576 77685-327
(R)-1-tert-Butyl-2-methyl pyrrolidine-1,2-dicarboxylate 97%
(R)-1-tert-Butyl-2-methyl pyrrolidine-1,2-dicarboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₉NO₄
Boiling Pt: 288.6 °C (760 mm Hg)
Flash Pt: 128.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00190828
CAS Number: 73323-65-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty