Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-Glu-OMe 95%

Supplier: AMBEED, INC
Warning

A372135-1G A372135-5G A372135-10G A372135-250MG A372135-25G A372135-5G A372135-1G
103826-496EA 22 USD InStock
103826-496 103837-514 76817-526 76817-528 76840-762 76840-764 77306-846
Boc-Glu-OMe 95%
Boc-Glu-OMe
Precaución: for Research Use Only


Formula: C₁₁H₁₉NO₆
Boiling Pt: 428.4 °C (760 mm Hg)
Melting Pt: 119…123 °C
Flash Pt: 212.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00076931
CAS Number: 72086-72-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty