Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Carnosic acid 97%

Supplier: AMBEED, INC
Warning

A489673-10MG A489673-25MG A489673-1MG A489673-1G A489673-25MG A489673-25G A489673-500G
103792-742EA 7 USD InStock
103792-742 103792-744 103828-208 76774-430 76843-236 76843-238 77626-978 77878-264
Carnosic acid 97%
Carnosic acid
Precaución: for Research Use Only


Formula: C₂₀H₂₈O₄
Storage Temperature: Freezer
MDL Number: MFCD02259459
CAS Number: 3650-09-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty