Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-1,2-Diaminocyclohexane 98%

Supplier: AMBEED, INC
Danger

A113207-1G A113207-5G A113207-100G A113207-25G A113207-500G A113207-1G A113207-5G
103798-190EA 5 USD InStock
103798-190 103824-028 76687-678 76687-680 76687-682 77310-852 77310-854
cis-1,2-Diaminocyclohexane 98%
cis-1,2-Diaminocyclohexane
Precaución: for Research Use Only


Formula: C₆H₁₄N₂
Boiling Pt: 193.6 °C (760 mm Hg)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00063746
CAS Number: 1436-59-5
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty