Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-6-bromoindole-2-carboxylate 98%

Supplier: AMBEED, INC
Warning

A196544-1G A196544-5G A196544-10G A196544-250MG A196544-1G A196544-25G A196544-5G
103816-856EA 24 USD InStock
103816-856 103847-470 77030-010 77030-012 77085-430 77085-432 77085-434
Ethyl-6-bromoindole-2-carboxylate 98%
Ethyl-6-bromoindole-2-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₀BrNO₂
MW: 268.11 g/mol
Boiling Pt: 394.7 °C (760 mm Hg)
Flash Pt: 192.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094942
CAS Number: 103858-53-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty