Ethyl-α-bromophenylacetate 95%
Supplier: AMBEED, INC
|
Danger
|
A157149-5G
A157149-10G
A157149-25G
A157149-5G
103843-172EA
41
USD
103843-172
76689-608
77300-236
77318-992
Ethyl-α-bromophenylacetate 95%
Ethyl α-bromophenylacetate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁BrO₂ MW: 243.10 g/mol Boiling Pt: 89…92 °C at 0.9 mmHg Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00013536 CAS Number: 2882-19-1 UN: 3265 ADR: 8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...